Order Entry
United States
ContactUsLinkComponent
 
Human TNF alpha ELISA Kit
Catalog # 77208-642
Supplier:  Avantor
CAS Number:  
undefined
Human TNF alpha ELISA Kit
Catalog # 77208-642
Supplier:  Avantor
CAS Number:  

Specifications

  • Assay duration:
    Multiple steps
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Rat
  • Target protein:
    TNF alpha
  • Description:
    Human TNF alpha ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Plasma, serum and lysates
  • Detection Method:
    Colorimetric
  • Time to Results:
    4 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    16 - 2000 pg/ml
  • Storage Temperature:
    4 °C
  • Regulatory Status:
    RUO
  • Cat. No.:
    77208-642

Specifications

About this item

This Human TNF alpha ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Human TNF alpha in plasma, serum, and lysates.