Order Entry
United States
Orders LinkContactUsLinkComponent
Mouse TGF beta 1 ELISA Kit

Mouse TGF beta 1 ELISA Kit

Mouse TGF beta 1 ELISA kit, 96 tests
Catalog # 77214-198
Mouse TGF beta 1 ELISA Kit
Catalog # 77214-198
Supplier:  
CAS Number:  
Mouse TGF beta 1 ELISA Kit
Mouse TGF beta 1 ELISA Kit

Mouse TGF beta 1 ELISA kit, 96 tests

CAS Number:  
Discontinued
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

This Mouse TGF beta 1 ELISA kit applies a technique called a quantitative sandwich immunoassay. It allows for the detection and quantification of endogenous levels of natural or recombinant Mouse TGF beta 1 in plasma, serum, and lysates.

Specifications

Specifications

  • Description:
    Mouse TGF beta 1 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    32 - 2000 pg/ml
  • Assay Type:
    Sandwich
  • Time to Results:
    4 h 30 min
  • Detection Method:
    Colorimetric
  • Cat. No.:
    77214-198
  • Regulatory Status:
    RUO
  • Assay duration:
    Multiple steps
  • Format:
    Pre-coated
  • Target protein:
    TGF beta 1
  • Sample Type:
    Plasma, serum and lysates
  • Shelf Life:
    Store for 6 months at 4 °C
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.