Order Entry
United States
ContactUsLinkComponent
L(+)-Sodium lactate solution ∼50%, EMPROVE® EXPERT Ph. Eur., BP, USP suitable for use as excipient, SAFC®
L(+)-Sodium lactate solution ∼50%, EMPROVE® EXPERT Ph. Eur., BP, USP suitable for use as excipient, SAFC®
Catalog # EM1.06522.2500
Supplier:  MilliporeSigma
CAS Number:  
L(+)-Sodium lactate solution ∼50%, EMPROVE® EXPERT Ph. Eur., BP, USP suitable for use as excipient, SAFC®
Catalog # EM1.06522.2500
Supplier:  MilliporeSigma
Supplier Number:  1.06522.2500
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    2,5 L
  • Cat. No.:
    EM1.06522.2500
  • Supplier no.:
    1.06522.2500

Specifications

Specification Test Results

  • Assay (Perchloric acid titration)#:
    50.0 - 51.0 %
  • Assay (Stereochemical purity of Sodium-(S)-lactate)#:
    ≥ 95.0 %
  • Identity (Sodium):
    passes test
  • Identity (Lactat):
    passes test
  • Appearance of solution:
    Clear and not more intense in color than reference solution BY₇
  • pH (in present substance):
    6.5 - 9.0
  • Chloride (Cl)#:
    ≤ 50 ppm
  • Sulfate (SO₄) (Ph Eur)#:
    ≤ 100 ppm
  • Sulfate (SO₄) (USP):
    passes test
  • Al (Aluminium):
    ≤ 0.1 ppm
  • As (Arsenic)#:
    ≤ 3 ppm
  • Fe (Iron)#:
    ≤ 10 ppm
  • Hg (Mercury):
    ≤ 1 ppm
  • Pb (Lead):
    ≤ 5 ppm
  • Oxalate and Phosphate (Ph Eur):
    passes test
  • Citrate, Oxalate, Phosphate or Tartrate (USP):
    passes test
  • Methanol and Methyl Esters:
    ≤ 250 ppm
  • Reducing sugars and sucrose (Ph Eur):
    passes test
  • Sugar (USP):
    passes test
  • Methanol#:
    ≤ 50 ppm
  • Residual solvents (ICH Q3C):
    excluded by manufacturing process
  • Total aerobic microbial count (TAMC):
    ≤ 100 CFU/ml
  • Total combined yeasts/moulds count (TYMC):
    ≤ 10 CFU/ml
  • Bile-tolerant gram-negative bacteria (absent in 1 ml):
    passes test
  • Candida albicans (absent in 1 ml):
    passes test
  • Escherichia coli (absent in 1 ml):
    passes test
  • Pseudomonas aeruginosa (absent in 1 ml):
    passes test
  • Salmonella (absent in 10 ml):
    passes test
  • Staphylococcus aureus (absent in 1 ml):
    passes test
  • Endotoxins:
    < 5

Specification Test Results

About this item

 :
 :112.06 g/mol
Boiling Pt:109 °C (1013 hPa)
Density:1.263 g/cm3 (20 °C)
 :M-Clarity™ quality level = MQ500 (more info.)
Storage Temperature:Ambient
CAS Number:867-56-1
Formula:C₃H₅NaO₃
MDL Number:MFCD00066576