Order Entry
United States
Orders LinkContactUsLinkComponent
Human LOXL2 ELISA Kit

Human LOXL2 ELISA Kit

Human LOXL2 ELISA kit, 96 tests
Catalog # 77173-890
Supplier:   Antibodies.com
Supplier #:  A313481-96
Human LOXL2 ELISA Kit
Catalog # 77173-890
Supplier:  Antibodies.com
CAS Number:  
Human LOXL2 ELISA Kit
Human LOXL2 ELISA Kit

Human LOXL2 ELISA kit, 96 tests

CAS Number:  
Price
$591.17
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Human LOXL2 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human LOXL2 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 150 to 2400 ng/L
  • Sensitivity: 9.35 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Human LOXL2 ELISA kit (A313481) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human LOXL2 in serum, plasma or other biological fluids. An antibody specific for LOXL2 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the LOXL2 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-LOXL2 antibody, which also binds the LOXL2 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-LOXL2 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-LOXL2 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of LOXL2 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of LOXL2 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.

Specifications

Specifications

  • Description:
    Human LOXL2 ELISA kit
  • Size:
    96 tests
  • Primary antibody reactivity:
    Human
  • Detection Range:
    150 - 2400 ng/L
  • Sample Volume:
    40 μl
  • Assay Type:
    Sandwich
  • Time to Results:
    1 h 30 min
  • Detection Method:
    Colorimetric
  • Cat. No.:
    77173-890
  • Regulatory Status:
    RUO
  • Assay duration:
    One step
  • Sensitivity:
    9.35 ng/L
  • Format:
    Pre-coated
  • Target protein:
    LOXL2
  • Sample Type:
    Serum, plasma or other biological fluids
  • Shelf Life:
    Store for 6 months at 4 °C
  • Storage Temperature:
    4 °C

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.