Order Entry
United States
ContactUsLinkComponent
 
Human IL-2RG ELISA Kit
  77174-320
 :  
undefined
Human IL-2RG ELISA Kit
  77174-320
 :  A313697-96
 :  

 

  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1.5 - 24 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.046 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77174-320

 

 

Human IL-2RG ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-2RG in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 1.5 to 24 ng/ml
  • Sensitivity: 0.046 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Human IL-2RG ELISA kit (A313697) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human IL-2RG in serum, plasma or other biological fluids. An antibody specific for IL-2RG has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-2RG present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-IL-2RG antibody, which also binds the IL-2RG present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-2RG antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-2RG antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of IL-2RG captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of IL-2RG in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

 : This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.