Order Entry
United States
ContactUsLinkComponent
Human IL-23R ELISA Kit
Human IL-23R ELISA Kit
Catalog # 77173-616
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human IL-23R ELISA Kit
Catalog # 77173-616
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A313343-96
CAS Number:  

Specifications

  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1.5 - 24 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.043 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77173-616

Specifications

About this item

Human IL-23R ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-23R in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 1.5 to 24 ng/ml
  • Sensitivity: 0.043 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Human IL-23R ELISA kit (A313343) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human IL-23R in serum, plasma or other biological fluids. An antibody specific for IL-23R has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-23R present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-IL-23R antibody, which also binds the IL-23R present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-23R antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-23R antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of IL-23R captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of IL-23R in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.