Order Entry
United States
ContactUsLinkComponent
Human IL-17RA Receptor ELISA Kit
Human IL-17RA Receptor ELISA Kit
Catalog # 77206-006
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human IL-17RA Receptor ELISA Kit
Catalog # 77206-006
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A311572-96
CAS Number:  

Specifications

  • Cat. No.:
    77206-006

Specifications

About this item

Human IL-17RA Receptor ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IL-17RA Receptor in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 300 to 4800 ng/L
  • Sensitivity: 16.15 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Human IL-17RA Receptor ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of human IL-17RA Receptor in serum, plasma or other biological fluids. An antibody specific for IL-17RA Receptor has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the IL-17RA Receptor present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-IL-17RA Receptor antibody, which also binds the IL-17RA Receptor present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-IL-17RA Receptor antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-IL-17RA Receptor antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of IL-17RA Receptor captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of IL-17RA Receptor in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.