Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Assays
ELISAs
Human IL-13 R Alpha 2 ELISA Kit
Human IL-13 R Alpha 2 ELISA Kit
Human IL-13 R Alpha 2 ELISA Kit
Catalog # 77212-070
Supplier:
Avantor
CAS Number:
Expand image
Human IL-13 R Alpha 2 ELISA Kit
Catalog # 77212-070
Supplier:
Avantor
CAS Number:
Specifications
Cat. No.:
77212-070
Specifications
About this item
Product Details
Product Details