Mouse IL-1 Beta ELISA Kit
Catalog # 77204-854
Supplier: Avantor
CAS Number:
Specifications
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:IL-1 beta
- Description:Mouse IL-1 beta ELISA kit
- Size:96 tests
- Detection Method:Chemiluminescent (Luminometer)
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:15 - 1000 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77204-854