Mouse IGF-1 ELISA Kit
77206-596
: Avantor
:
- Assay duration:Multiple steps
- Assay Type:Sandwich
- Format:Pre-coated
- Primary antibody reactivity:Mouse
- Target protein:IGF-1
- Description:Mouse IGF-1 ELISA kit
- Size:96 tests
- Detection Method:Chemiluminescent (Luminometer)
- Shelf Life:Store for 6 months at 4 °C
- Detection Range:23 - 23 - 1500 pg/ml
- Storage Temperature:4 °C
- Regulatory Status:RUO
- Cat. No.:77206-596