Order Entry
United States
ContactUsLinkComponent
Human ICAM3 ELISA Kit
Human ICAM3 ELISA Kit
Catalog # 77173-302
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human ICAM3 ELISA Kit
Catalog # 77173-302
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A313185-96
CAS Number:  

Specifications

  • Cat. No.:
    77173-302

Specifications

About this item

Human ICAM3 ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human ICAM3 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 15 to 240 ng/ml
  • Sensitivity: 0.41 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra to Assay: CV <8%, Inter to Assay: CV <10%

Human ICAM3 ELISA kit (A313185) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human ICAM3 in serum, plasma or other biological fluids. An antibody specific for ICAM3 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the ICAM3 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-ICAM3 antibody, which also binds the ICAM3 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-ICAM3 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-ICAM3 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of ICAM3 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of ICAM3 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.