Order Entry
United States
ContactUsLinkComponent
Mouse GDNF ELISA Kit
Mouse GDNF ELISA Kit
Catalog # 77212-208
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse GDNF ELISA Kit
Catalog # 77212-208
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A311537-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    GDNF
  • Description:
    Mouse GDNF ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    4 - 64 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.213 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77212-208

Specifications

About this item

Mouse GDNF ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse GDNF in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 4 to 64 ng/L
  • Sensitivity: 0.213 ng/L
  • Sample Type: Serum, plasma or other biological fluids
  • Assay precision: Intra assay: CV <8%, Inter assay: CV <10%

Mouse GDNF ELISA kit employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse GDNF in serum, plasma or other biological fluids. An antibody specific for GDNF has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the GDNF present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-GDNF antibody, which also binds the GDNF present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-GDNF antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-GDNF antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of GDNF captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of GDNF in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready to use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.