Order Entry
United States
ContactUsLinkComponent
Human FGL2/Prothrombinase ELISA Kit
Human FGL2/Prothrombinase ELISA Kit
Catalog # 77175-492
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Human FGL2/Prothrombinase ELISA Kit
Catalog # 77175-492
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A314287-96
CAS Number:  

Specifications

  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    1.5 - 24 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.058 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77175-492

Specifications

About this item

Human FGL2/Prothrombinase ELISA kit is a 90 minutes sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human FGL2/Prothrombinase in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutess, with a single wash step
  • Detection Range: 1.5 - 24 ng/ml
  • Sensitivity: 0.058 ng/ml
  • Sample Type: Serum, plasma or other biological fluids
  • Assay Precision: Intra - Assay: CV <8%, Inter - Assay: CV <10%

Human FGL2/Prothrombinase ELISA kit (A314287) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human FGL2/Prothrombinase in serum, plasma or other biological fluids. An antibody specific for FGL2/Prothrombinase has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the FGL2/Prothrombinase present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-FGL2/Prothrombinase antibody, which also binds the FGL2/Prothrombinase present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-FGL2/Prothrombinase antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-FGL2/Prothrombinase antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of FGL2/Prothrombinase captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of FGL2/Prothrombinase in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, standard solution, standard diluent, biotinylated detection antibody, steptavidin-HRP, wash buffer (25X), substrate solution A, substrate solution B, stop solution, plate sealers, foil pouch, and protocol manual.