Order Entry
United States
ContactUsLinkComponent
Mouse Angiotensin Converting Enzyme 1 ELISA Kit
Mouse Angiotensin Converting Enzyme 1 ELISA Kit
Catalog # 77213-486
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
Mouse Angiotensin Converting Enzyme 1 ELISA Kit
Catalog # 77213-486
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A312598-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Angiotensin Converting Enzyme 1
  • Description:
    Mouse angiotensin converting enzyme 1 ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    7.5 - 120 ng/ml
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    0.28 ng/ml
  • Regulatory Status:
    RUO
  • Cat. No.:
    77213-486

Specifications

About this item

Mouse Angiotensin Converting Enzyme 1 ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Angiotensin Converting Enzyme 1 in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Angiotensin Converting Enzyme 1 ELISA kit (A312598) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Angiotensin Converting Enzyme 1 in serum, plasma or other biological fluids. An antibody specific for Angiotensin Converting Enzyme 1 has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Angiotensin Converting Enzyme 1 present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Angiotensin Converting Enzyme 1 antibody, which also binds the Angiotensin Converting Enzyme 1 present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Angiotensin Converting Enzyme 1 antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Angiotensin Converting Enzyme 1 antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Angiotensin Converting Enzyme 1 captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Angiotensin Converting Enzyme 1 in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.