Order Entry
United States
ContactUsLinkComponent
Mouse Activin Receptor Type IIB/ACVR2B ELISA Kit
Mouse Activin Receptor Type IIB/ACVR2B ELISA Kit
Catalog # 77173-068
Supplier:  ANTIBODIES.COM LLC
CAS Number:  
undefined
Mouse Activin Receptor Type IIB/ACVR2B ELISA Kit
Catalog # 77173-068
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A313066-96
CAS Number:  

Specifications

  • Assay duration:
    One step
  • Assay Type:
    Sandwich
  • Format:
    Pre-coated
  • Primary antibody reactivity:
    Mouse
  • Target protein:
    Activin Receptor Type IIB/ACVR2B
  • Description:
    Mouse activin receptor type IIB/ACVR2B ELISA kit
  • Size:
    96 tests
  • Sample Type:
    Serum, plasma or other biological fluids
  • Detection Method:
    Colorimetric
  • Time to Results:
    1 h 30 min
  • Shelf Life:
    Store for 6 months at 4 °C
  • Detection Range:
    75 - 1200 ng/L
  • Storage Temperature:
    4 °C
  • Sample Volume:
    40 μl
  • Sensitivity:
    5.48 ng/L
  • Regulatory Status:
    RUO
  • Cat. No.:
    77173-068

Specifications

About this item

Mouse Activin receptor Type IIB/ACVR2B ELISA kit is a 90 minute sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of mouse Activin receptor Type IIB/ACVR2B in serum, plasma, and other biological fluids.

  • Higher throughput: Get results in just 90 minutes, with a single wash step
  • Assay precision: Intra-assay: CV <8%, Inter-assay: CV <10%

Mouse Activin receptor Type IIB/ACVR2B ELISA kit (A313066) employs the sandwich enzyme immunoassay technique for the quantitative measurement of mouse Activin receptor Type IIB/ACVR2B in serum, plasma or other biological fluids. An antibody specific for Activin receptor Type IIB/ACVR2B has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the Activin receptor Type IIB/ACVR2B present in each sample is bound to the wells by the immobilized antibody. Biotinylated anti-Activin receptor Type IIB/ACVR2B antibody, which also binds the Activin receptor Type IIB/ACVR2B present in each sample, and Streptavidin-HRP, which binds the Biotinylated anti-Activin receptor Type IIB/ACVR2B antibody, are added and the microtiter plate is incubated. Following incubation, unbound Biotinylated anti-Activin receptor Type IIB/ACVR2B antibody and unbound Streptavidin-HRP are removed by washing, and two substrate solutions are added to the wells. Color develops in proportion to the amount of Activin receptor Type IIB/ACVR2B captured in each well. The color development is stopped by addition of stop solution which changes the color from blue to yellow and the intensity of the color is then measured. The concentration of Activin receptor Type IIB/ACVR2B in the samples can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.

Ordering information: This is a ready-to-use ELISA kit that contains: Pre-coated 96 well microplate, positive control, negative control, HRP-conjugated detection antibody, sample diluent, substrate solution A, substrate solution B, stop solution, wash buffer (25X), plate sealers, foil pouch, and protocol manual.