ContactUsLinkComponent
PureAmp™ Sep-R-8™ PCR Tube Strips
PureAmp™ Sep-R-8™ PCR Tube Strips
Catalog # 470353-202
Supplier:  MTC BIO, INC.
undefined
PureAmp™ Sep-R-8™ PCR Tube Strips
Catalog # 470353-202
Supplier:  MTC BIO, INC.
Supplier Number:  P9602-N-T
Specifications
  • Description:
    PureAmp™ Sep-R-8™ PCR tube strips
  • Cat. No.:
    470353-202
Specifications
About this item
These PCR plates can be broken down into strips of eight tubes for greater control over sample quantity, and to ensure reduced waste.
  • RNase, DNase, DNA, and PCR inhibitor free
  • Compatible with standard equipment
  • Ultra-thin walled tube wells
  • Alpha-numeric reference grids
All PureAmp™ PCR plates are free of RNase, DNase, DNA, and PCR inhibitors, and are made of heat-resistant, medical-grade polypropylene.