Order Entry
United States
ContactUsLinkComponent
Anti-Rabies Human Recombinant Antibody [clone: RV30]
Anti-Rabies Human Recombinant Antibody [clone: RV30]
Supplier:  ReVacc Scientific
Certificates
undefined
Anti-Rabies Human Recombinant Antibody [clone: RV30]
Supplier:  ReVacc Scientific

Specifications

  • Catalog No:
  • 77257-178
  • 77257-180
  • Antigen Symbol:
  • Rabies Virus
  • Rabies Virus
  • Antigen Name:
  • Glycoprotein
  • Glycoprotein
  • Antigen Synonyms:
  • GP
  • GP
  • Antibody Type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • RV30
  • RV30
  • Reactivity:
  • Human
  • Human
  • Host:
  • Human
  • Human
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western Blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Flow Cytometry:
  • Yes
  • Yes
  • Neutralisation:
  • Yes
  • Yes
  • Modification:
  • Recombinant monoclonal antibody
  • Recombinant monoclonal antibody
  • Molecular Weight:
  • 150 kDa
  • 150 kDa
  • Purification:
  • Purified by Protein A chromatography
  • Purified by Protein A chromatography
  • Concentration:
  • 1 mg/ml by refractometry
  • 1 mg/ml by refractometry
  • Storage Buffer:
  • PBS, pH 7.2
  • PBS, pH 7.2
  • Storage Temperature:
  • Store at 4 °C, −20 °C
  • Store at 4 °C, −20 °C
  • Shipping Temperature:
  • Ice
  • Ice

Specifications

Frequently Bought Together