Anti-PIP5K1A Rabbit Polyclonal Antibody
Catalog # 76756-618
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha
- Antigen Symbol:PIP5K1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:8394
- Antigen Synonyms:PtdIns(4)P-5-kinase 1 alpha|PIP5K1-alpha|68 kDa type I phosphatidylinositol 4-phosphate 5-kinase alpha|Phosphatidylinositol 4-phosphate 5-kinase type I alpha|Phosphatidylinositol 4-phosphate 5-kinase type-1 alpha|PIP5K1A|PIP5KIalpha
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 231-500 of human PIP5K1A (NP_001129109.1).
- Purification:Affinity purification
- Cat. No.:76756-618
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PIP5K1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse