Anti-GNB1L Rabbit Polyclonal Antibody
Catalog # 76753-180
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Guanine nucleotide binding protein (G protein), beta polypeptide 1-like
- Antigen Symbol:GNB1L
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:54584
- Antigen Synonyms:WDVCF|GY2|FKSG1|DGCRK3|WDR14|GNB1L
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-327 of human GNB1L (NP_443730.1).
- Purification:Affinity purification
- Cat. No.:76753-180
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: GNB1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat