Anti-RASSF5 Rabbit Polyclonal Antibody
Catalog # 76753-146
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Ras association (RalGDS/AF-6) domain family member 5
- Antigen Symbol:RASSF5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:83593
- Antigen Synonyms:NORE1A|New ras effector 1|Maxp1|Ras association domain-containing protein 5|NORE1|NORE1B|RASSF3|RAPL
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-265 of human RASSF5 (NP_872606.1).
- Purification:Affinity purification
- Cat. No.:76753-146
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: RASSF5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat