Anti-SMYD1 Rabbit Polyclonal Antibody
76752-930
: Prosci
- Antibody Type:Primary
- Antigen Name:Histone-lysine N-methyltransferase SMYD1
- Antigen Symbol:SMYD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:150572
- Antigen Synonyms:SET and MYND domain-containing protein 1|SMYD1|Histone-lysine N-methyltransferase SMYD1
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-350 of human SMYD1 (NP_938015.1).
- Purification:Affinity purification
- Cat. No.:76752-930
- Packaging:Vial
Type: Primary
Antigen: SMYD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat