Anti-SMC1A Rabbit Polyclonal Antibody
Catalog # 76759-042
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Structural maintenance of chromosomes 1A
- Antigen Symbol:SMC1A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:8243
- Antigen Synonyms:SMC-1-alpha|SMC1|DXS423E|SMC-1A|CDLS2|SB1.8|SMC1A|KIAA0178|SMC protein 1A|SMC1L1|SMCB
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human SMC1A (NP_006297.2).
- Purification:Affinity purification
- Cat. No.:76759-042
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SMC1A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat