Anti-HNRNPM Rabbit Polyclonal Antibody
76762-560
: Prosci
- Antibody Type:Primary
- Antigen Name:Heterogeneous nuclear ribonucleoprotein M
- Antigen Symbol:HNRNPM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:4670
- Antigen Synonyms:HNRNPM4|HNRPM4|DKFZp547H118|HNRPM|HNRNPM|NAGR1|CEAR|HTGR1|hnRNP M
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 491-730 of human HNRNPM (NP_005959.2).
- Purification:Affinity purification
- Cat. No.:76762-560
- Packaging:Vial
Type: Primary
Antigen: HNRNPM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat