Anti-COL10A1 Rabbit Polyclonal Antibody
Catalog # 76756-058
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Collagen alpha-1(X) chain
- Antigen Symbol:COL10A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:50 µl
- Form:Liquid
- Gene ID:1300
- Antigen Synonyms:COL10A1|Collagen alpha-1(X) chain
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 521-680 of human COL10A1 (NP_000484.2).
- Purification:Affinity purification
- Cat. No.:76756-058
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: COL10A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat