Anti-PUF60 Rabbit Polyclonal Antibody
Catalog # 76758-060
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Poly-U binding splicing factor 60KDa
- Antigen Symbol:PUF60
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:22827
- Antigen Synonyms:SIAHBP1|FLJ31379|PUF60|FIR|VRJS|RoBPI
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 243-542 of human PUF60 (NP_055096.2).
- Purification:Affinity purification
- Cat. No.:76758-060
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: PUF60
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat