Anti-HAS3 Rabbit Polyclonal Antibody
Catalog # 76762-376
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Hyaluronan synthase 3
- Antigen Symbol:HAS3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3038
- Antigen Synonyms:Hyaluronic acid synthase 3|Hyaluronate synthase 3|HAS3|Hyaluronan synthase 3|HA synthase 3
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 67-281 of human HAS3 (NP_619515.1).
- Purification:Affinity purification
- Cat. No.:76762-376
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HAS3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat