Anti-CPM Rabbit Polyclonal Antibody
Catalog # 76751-586
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Carboxypeptidase M
- Antigen Symbol:CPM
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:1368
- Antigen Synonyms:Carboxypeptidase M|CPM
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 204-443 of human CPM (NP_001005502.1).
- Purification:Affinity purification
- Cat. No.:76751-586
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CPM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human