Anti-CNOT1 Rabbit Polyclonal Antibody
Catalog # 76750-854
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CCR4-NOT transcription complex subunit 1
- Antigen Symbol:CNOT1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:23019
- Antigen Synonyms:CDC39|NOT1 (negative regulator of transcription 1|NOT1H|yeast) homolog|CCR4-NOT transcription complex subunit 1|negative regulator of transcription subunit 1 homolog|AD-005|adrenal gland protein AD-005|NOT1|CCR4-associated factor 1
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 2137-2376 of human CNOT1 (NP_057368.3).
- Purification:Affinity purification
- Cat. No.:76750-854
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CNOT1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human