Anti-CRX Rabbit Polyclonal Antibody
Catalog # 76750-498
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Cone-rod homeobox
- Antigen Symbol:CRX
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:1406
- Antigen Synonyms:CRX|LCA7|OTX3|cone-rod homeobox protein|CRD|orthodenticle homeobox 3|CORD2|cone-rod homeobox
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 120-285 of human CRX (NP_000545.1).
- Purification:Affinity purification
- Cat. No.:76750-498
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CRX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat