Anti-LBR Rabbit Polyclonal Antibody
Catalog # 76763-832
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Lamin B receptor
- Antigen Symbol:LBR
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3930
- Antigen Synonyms:delta(14)-sterol reductase|PHA|TDRD18|delta-14-SR|LMN2R|DHCR14B|3-beta-hydroxysterol Delta (14)-reductase|PHASK|C-14 sterol reductase|C14SR|integral nuclear envelope inner membrane protein|tudor domain containing 18|sterol C14-reductase
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-211 of human LBR (NP_002287.2).
- Purification:Affinity purification
- Cat. No.:76763-832
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: LBR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat