Anti-EEF2K Rabbit Polyclonal Antibody
Catalog # 76757-318
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Eukaryotic elongation factor 2 kinase
- Antigen Symbol:EEF2K
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:29904
- Antigen Synonyms:alternative protein EEF2K|elongation factor-2 kinase|eukaroytic elongation factor 2 kinase|eukaryotic elongation factor 2 kinase|CaMKIII|eEF-2 kinase|calcium/calmodulin-dependent eukaryotic elongation factor-2 kinase|HSU9385eEF-2K|calmodulin-dependent protein kinase III
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 406-725 of human EEF2K (NP_037434.1).
- Purification:Affinity purification
- Cat. No.:76757-318
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: EEF2K
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse