Anti-SERPINA10 Rabbit Polyclonal Antibody
Catalog # 76756-682
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 10
- Antigen Symbol:SERPINA10
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:51156
- Antigen Synonyms:protein Z-dependent protease inhibitor|clade A (alph|antitrypsin)|ZPI|serpin peptidase inhibitor|serine (or cysteine) proteinase inhibitor|PZI|SERPINA10|clade A (alpha-1 antiproteinase|protein Z-dependent protease inhibitorprecursor|member 10|UNQ707/PRO1358
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 165-444 of human SERPINA10 (NP_001094077.1).
- Purification:Affinity purification
- Cat. No.:76756-682
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SERPINA10
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat