Anti-CTR9 Rabbit Polyclonal Antibody
Catalog # 76764-308
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CTR9 homolog, Paf1/RNA polymerase II complex component
- Antigen Symbol:CTR9
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:9646
- Antigen Synonyms:homolog|SH2-binding phosphoprotein|SH2 domain binding protein 1 (tetratricopeptide repeat containing)|RNA polymerase-associated protein CTR9 homolog|SH2BP1|Paf1/RNA polymerase II complex component|p15p150TSP|Ctr9|TSBP|TPR-containing
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1024-1173 of human CTR9 (NP_055448.1).
- Purification:Affinity purification
- Cat. No.:76764-308
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CTR9
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat