Anti-DNMT3B Rabbit Polyclonal Antibody
Catalog # 76757-086
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:DNA (cytosine-5)-methyltransferase 3B
- Antigen Symbol:DNMT3B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:1789
- Antigen Synonyms:DNA MTase HsaIIIB|DNA (cytosine-5)-methyltransferase 3B|Dnmt3b|MHsaIIIB|DNA methyltransferase HsaIIIB|DNMT3B
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at −20...+730 °C. Avoid freeze/thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:A synthetic peptide of human DNMT3B.
- Purification:Affinity purification
- Cat. No.:76757-086
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: DNMT3B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat