Human Recombinant B7-H5 Protein (from HEK293 Cells)
Catalog # 77134-454
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:HEK293 cells
- Species:Human
- Size:0.1 mg
- Storage Conditions:Lyophilized Protein should be stored at −20 °C or lower for long term storage. Upon reconstitution, working aliquots should be stored at −20 or −70 °C. Avoid repeated freeze-thaw cycles.
- Biological Activity:Measured by its binding ability in a functional ELISA. Immobilized Human VSIG8, His Tag at 10 µg/ml (100 µl/well) can bind Human B7-H5, Fc Tag with a linear range of 0.625 - 20 µg/ml.
- Gene ID:64115
- Protein Synonyms:B7-H5|SISP1|Gi24|VISTA
- Protein/Peptide Name:B7-H5
- Purity:>95% as determined by SDS-PAGE. Endotoxin level is less than 1.0 EU per µg by the LAL method.
- Molecular Weight:44.8 kDa
- Sequence:Phe 33 - Ala 194
- Formulation:Lyophilized. PBS, pH 7.4.
- Shipping Temperature:RT
- Tested Applications:ELISA, WB
- Cat. No.:77134-454