SARS-CoV-2 (COVID-19) Envelope Peptide
77126-030
: Prosci
:
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Source:
- Species:SARS-CoV-2
- Size:0.05 mg
- Environmentally Preferable:
- Storage Conditions:Store SARS-CoV-2 (COVID-19) Envelope Peptide at −20 °C, stable for one year.
- Gene ID:43740570
- Protein Synonyms:E protein|Envelope protein|SARS-CoV-2 (COVID-19|2019-nCoV) Envelope Antibody: Severe acute respiratory syndrome coronavirus 2 (SARS-CoV-2)
- Protein/Peptide Name:Envelope
- Concentration:200 µg/ml
- Formulation:Liquid. PBS pH 7.2 (10 mM NaH₂PO₄, 10 mM Na₂HPO₄, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide.
- Shipping Temperature:20
- Tested Applications:Blocking
- Cat. No.:77126-030