His-tag Peptide
Catalog # 77120-572
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Synthetic
- Species:Human
- Size:0.05 mg
- Storage Conditions:Store His-tag peptide at −20 °C, stable for one year.
- Protein Synonyms:6xHis|His-tag|His
- Concentration:200 µg/ml
- Formulation:Liquid. PBS pH 7.2 (10 mM NaH₂PO₄, 10 mM Na₂HPO₄, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide.
- Shipping Temperature:20
- Tested Applications:Blocking
- Cat. No.:77120-572