Order Entry
United States
ContactUsLinkComponent
 
PHC3 Peptide
  77123-350
 :  Prosci
 :  
PHC3 Peptide
  77123-350
 :  Prosci
 :  7607P
 :  

 

  • Pack type:
    Vial
  • Conjugation:
    Unconjugated
  • Protein/Peptide Type:
    Synthetic
  • Species:
    Human
  • Size:
    0.05 mg
  • Storage Conditions:
    Store PHC3 peptide at −20 °C, stable for one year.
  • Gene ID:
    80012
  • Protein Synonyms:
    PH3|hPH3|Polyhomeotic-like protein 3|EDR3|HPH3|Early development regulatory protein 3
  • Protein/Peptide Name:
    PHC3
  • Concentration:
    200 µg/ml
  • Formulation:
    Liquid. PBS pH 7.2 (10 mM NaH₂PO₄, 10 mM Na₂HPO₄, 130 mM NaCl) containing 0.1% bovine serum albumin and 0.02% sodium azide.
  • Shipping Temperature:
    20
  • Tested Applications:
    Blocking
  • Cat. No.:
    77123-350