Rubella Virus Recombinant Envelope 1 Protein (from E. coli)
Catalog # 77124-796
Supplier: Prosci
CAS Number:
Specifications
- Pack type:Vial
- Conjugation:Unconjugated
- Protein/Peptide Type:Recombinant
- Source:E. coli
- Species:Rubella
- Size:0.01 mg
- Storage Conditions:Store in working aliquots at −20 °C. Avoid freeze/thaw cycles. When working with proteins care should be taken to keep recombinant protein at a cool and stable temperature.
- Protein/Peptide Name:E1
- Purity:~95% by SDS-PAGE
- Molecular Weight:66 kDa
- Concentration:Batch dependent
- Formulation:Liquid. 25 mM Tris-HCl, 0.1% SDS, pH 8.3.
- Shipping Temperature:20
- Tested Applications:ELISA
- Cat. No.:77124-796