Anti-ACTL7B Rabbit Polyclonal Antibody
Catalog # 76763-454
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:actin-like 7B
- Antigen Symbol:ACTL7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10880
- Antigen Synonyms:RP11-3J11.2|actin-like 7-beta|actin-like 7B|Tact1|ACTL7B
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 200-380 of human ACTL7B (NP_006677.1).
- Purification:Affinity purification.
- Cat. No.:76763-454
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ACTL7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat