Anti-SULT1A1 Rabbit Polyclonal Antibody
Catalog # 76749-534
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Sulfotransferase 1A1
- Antigen Symbol:SULT1A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:6817
- Antigen Synonyms:Thermostable phenol sulfotransferase|Ts-PST|ST1A1|P-PST 1|STP1|Phenol-sulfating phenol sulfotransferase 1|STP|SULT1A1|ST1A3|HAST1/HAST2|Phenol sulfotransferase 1|Aryl sulfotransferase 1|Sulfotransferase 1A1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-295 of human SULT1A1 (NP_803565.1).
- Purification:Affinity purification.
- Cat. No.:76749-534
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SULT1A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human