Anti-VEGFA Rabbit Polyclonal Antibody
Catalog # 76764-450
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:vascular endothelial growth factor A
- Antigen Symbol:VEGFA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:7422
- Antigen Synonyms:VEGF|VEGFA|VEGF-A|Vascular permeability factor|VPF|Vascular endothelial growth factor A
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 27-126AA of human VEGF (NP_001165099.1).
- Purification:Affinity purification.
- Cat. No.:76764-450
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: VEGFA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat