Anti-CD300C Rabbit Polyclonal Antibody
Catalog # 76752-232
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:CMRF35-like molecule 6
- Antigen Symbol:CD300C
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:10871
- Antigen Synonyms:CMRF35|CLM-6|CMRF35-A1|Immunoglobulin superfamily member 16|CMRF35-like molecule 6|IgSF16|CMRF-35|CD300c|CD300C|CMRF35A|CMRF35A1|IGSF16|CD300 antigen-like family member C
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 21-183 of human CD300C (NP_006669.1).
- Purification:Affinity purification.
- Cat. No.:76752-232
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CD300C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat