Anti-ASPN Rabbit Polyclonal Antibody
Catalog # 76762-230
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:asporin
- Antigen Symbol:ASPN
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:54829
- Antigen Synonyms:ASPN|OS3|FLJ20129|SLRR1C|PLAP-1|PLAP1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 100-379 of human ASPN (NP_060150.4).
- Purification:Affinity purification.
- Cat. No.:76762-230
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: ASPN
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat