Anti-SLC23A1 Rabbit Polyclonal Antibody
Catalog # 76761-860
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:solute carrier family 23 member 1
- Antigen Symbol:SLC23A1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:9963
- Antigen Synonyms:YSPL3|yolk sac permease-like molecule 3|sodium-dependent vitamin C transporter-1|solute carrier family 23 (ascorbic acid transporter)|member 2|hSVCT1|Na(+)/L-ascorbic acid transporter 1|member 1|solute carrier family 23 (nucleobase transporters)|SLC23A2|SVCT1|solute carrier family 23 member 1
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 512-598 of human SLC23A1 (NP_005838.3).
- Purification:Affinity purification.
- Cat. No.:76761-860
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: SLC23A1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human