Anti-CCNK Rabbit Polyclonal Antibody
Catalog # 76763-778
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:Cyclin-K
- Antigen Symbol:CCNK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:8812
- Antigen Synonyms:CCNK|Cyclin-K|CPR4
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-160 of human CCNK (NP_001092872.1).
- Purification:Affinity purification.
- Cat. No.:76763-778
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: CCNK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat