Anti-IARS Rabbit Polyclonal Antibody
Catalog # 76754-814
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:isoleucyl-tRNA synthetase
- Antigen Symbol:IARS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3376
- Antigen Synonyms:isoleucine tRNA ligase 1|IRS|cytoplasmic|GRIDHH1|PRO0785|ILERS|isoleucine--tRNA ligase|IARS|ILRS
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1158-1262 of human IARS (NP_002152.2).
- Purification:Affinity purification.
- Cat. No.:76754-814
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: IARS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat