Anti-HK2 Rabbit Polyclonal Antibody
Catalog # 76749-818
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:hexokinase 2
- Antigen Symbol:HK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:50 µl
- Form:Liquid
- Gene ID:3099
- Antigen Synonyms:HK2|HKII|HXK2|DKFZp686M1669
- Modification:None
- Storage Buffer:PBS with 0.02% sodium azide, 50% glycerol, pH 7.3.
- Storage Temperature:Store at –20...+730 °C. Avoid freeze / thaw cycles.
- Concentration:Batch dependent
- Shipping Temperature:20
- Immunogen:Recombinant fusion protein containing a sequence corresponding to amino acids 1-120 of human Hexokinase II (NP_000180.2).
- Purification:Affinity purification.
- Cat. No.:76749-818
- Packaging:Vial
Specifications
About this item
Type: Primary
Antigen: HK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat