Anti-PGAM5 Rabbit Polyclonal Antibody
77125-432
: Prosci
- Antibody Type:Primary
- Antigen Name:PGAM family member 5, mitochondrial serine/threonine protein phosphatase
- Antigen Symbol:PGAM5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.02 mg
- Form:Liquid
- Gene ID:192111
- Antigen Synonyms:Bcl-XL-binding protein v68|Serine/threonine-protein phosphatase PGAM5 mitochondrial|Phosphoglycerate mutase family member 5
- Modification:None
- Storage Buffer:PGAM5 Antibody is supplied in PBS containing 0.02% sodium azide.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:Anti-PGAM5 antibody 9517 was raised against a peptide corresponding to 14 amino acids near the carboxyl terminus of human PGAM5. The immunogen is located within the last 50 amino acids of PGAM5.
- Purification:PGAM5 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:77125-432
- Packaging:Vial
PGAM5 antibody can be stored at 4 to 730 °C for three months and –20 to 730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type: Primary
Antigen: PGAM5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat