Anti-SIGLEC15 Rabbit Polyclonal Antibody
Catalog # 77127-588
Supplier: Prosci
Specifications
- Antibody Type:Primary
- Antigen Name:sialic acid binding Ig-like lectin 15
- Antigen Symbol:SIGLEC15
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:0.1 mg
- Form:Liquid
- Gene ID:284266
- Antigen Synonyms:Siglec-15|SIGLEC-15|SIGLEC15 Antibody: CD33L3|CD33L3|CD33 antigen-like 3|Sialic acid-binding Ig-like lectin 15|HsT1361
- Modification:None
- Storage Buffer:SIGLEC15 Antibody is supplied in PBS containing 0.02% sodium azide.
- Concentration:1 mg/ml
- Shipping Temperature:20
- Immunogen:Anti-SIGLEC15 antibody 9017 was raised against a peptide corresponding to 14 amino acids near the carboxy terminus of human SIGLEC15. The immunogen is located within amino acids 270-320 of SIGLEC15.
- Purification:SIGLEC15 Antibody is affinity chromatography purified via peptide column.
- Cat. No.:77127-588
- Packaging:Vial
Specifications
About this item
SIGLEC15 antibody can be stored at 4 to 730 °C for three months and –20 to +730 °C, stable for up to one year. As with all antibodies care should be taken to avoid repeated freeze thaw cycles. Antibodies should not be exposed to prolonged high temperatures.
Type: Primary
Antigen: SIGLEC15
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat